2,6-Di-O-methyl-β-cyclodextrine content > 35% (DS~14)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51166-71-3
:
Formula:C₅₆H₉₈O₃₅
MDL Number:MFCD00011616
:1331.3563 g/mol
Storage Temperature:Ambient
Frequently Bought Together