(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:124655-17-0
:
Formula:C₁₁H₂₁NO₄
MDL Number:MFCD00065575
:231.29 g/mol
Storage Temperature:Ambient
Frequently Bought Together