
4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:942518-20-9
:
Formula:C₁₉H₁₈N₄O₄
MDL Number:MFCD11052920
:366.38 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together