Order Entry
Netherlands
ContactUsLinkComponent
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamide)
Irritant
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamide)
Catalog # APOSBIB1157-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]cholamide)
Catalog # APOSBIB1157-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:86303-22-2
Formula:C₄₂H₇₅N₃O₁₆
MDL Number:MFCD00082541