
N-Acetyl-L-cysteine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:616-91-1
:
Formula:C₅H₉NO₃S
MDL Number:MFCD00004880
:163.2 g/mol
Melting Pt:106…111 °C
EINECS:210-498-3
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together