Human recombinant B7-H3 (from HEK293)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 at 2 μg/ml (100 μl/well) can bind Biotinylated Human B7-H3, Fc,Avitag with a linear range of 0,3-2 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Biotinylated Human B7-H3, Fc,Avitag at 1 μg/ml (100 μl/well) on Streptavidin precoated (0,5 μg/well) plate, can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0,3-5 ng/ml (Routinely tested)
- Gene ID:Q5ZPR3-2
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:B7 homolog 3|B7-H3|CD276
- Protein/peptide name:B7-H3
- Purity:>95%
- Molecular weight:51,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together