Order Entry
Netherlands
ContactUsLinkComponent
 
Mouse recombinant Siglec-10 (from HEK293 cells)
Catalog # ACRBSI0-M52H7-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Mouse recombinant Siglec-10 (from HEK293 cells)
Catalog # ACRBSI0-M52H7-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    Q80ZE3-1
  • Protein synonyms:
    Siglec10|PRO940|SIGLEC10|SLG2|MGC126774
  • Protein/peptide name:
    Siglec-10
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    60,7 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 25 mM MES, 150 mM NaCl, pH 5,5. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

About this item

Mouse Siglec-10 protein, His Tag.