Order Entry
Netherlands
ContactUsLinkComponent
Mouse recombinant nectin-4 (from HEK293 cells)
Mouse recombinant nectin-4 (from HEK293 cells)
Catalog # ACRBNE4-M52H3100UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Mouse recombinant nectin-4 (from HEK293 cells)
Catalog # ACRBNE4-M52H3100UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Mouse Nectin-4, His Tag (Cat. No. NE4-M52H3) at 5 μg/ml (100 μl/well) can bind Human PVRL1 / NECTIN1 Protein, Fc Tag with a linear range of 0,039 - 1,25 μg/ml (QC tested). Loaded Mouse Nectin-4, His Tag (Cat. No. NE4-M52H3) on HIS1K Biosensor, can bind Human Nectin-1, Fc Tag (Cat. No. PV1-H5253) with an affinity constant of 0,12 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q8R007-1
  • Protein synonyms:
    NECTIN4|PRR4|PVRL4|LNIR
  • Protein/peptide name:
    Nectin-4
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    36,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, Arginine, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

About this item

Mouse Nectin-4 protein, His Tag.