Human recombinant AIMP1 (from E. coli)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Gene ID:Q12904-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:EMAP-2|HLD3|p43|EMAPII|AIMP1|AIMP-1|EMAP-II|SCYE1|EMAP2
- Protein/peptide name:AIMP1
- Purity:>95%
- Molecular weight:35,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together