Human recombinant 4-1BB (from HEK293)
Catalog # ACRB41B-H5256-1MG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pack type:1 mg
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human 4‑1BB Ligand, Fc Tag at 1 μg/ml (100 μl/well) can bind Human 4-1BB, Mouse IgG2a Fc Tag, low endotoxin with a linear range of 0,6-5 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human 4‑1BB Ligand, His Tag at 2 μg/ml (100 μl/well) can bind Human 4-1BB, Mouse IgG2a Fc Tag, low endotoxin with a linear range of 0,1-4 ng/ml (Routinely tested).; Flow Cytometry assay shows that Human 4-1BB, Mouse IgG2a Fc Tag, low endotoxin can bind to 293T cells overexpressing Human 4-1BBL. The concentration of 4-1BB used is 0,03 μg/ml (Routinely tested)
- Gene ID:Q07011-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD137|TNFRSF9|ILA|CDw137|4-1BB
- Protein/peptide name:4-1BB
- Purity:>95%
- Molecular weight:44,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder,RT