Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant 2B4 (from HEK293)
Human recombinant 2B4 (from HEK293)
Catalog # ACRB2B4-H82F0-25UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human recombinant 2B4 (from HEK293)
Catalog # ACRB2B4-H82F0-25UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pack type:
    25 µg
  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA, Immobilised Human CD48, Fc tag at 10 μg/ml (100 μl/well) can bind Biotinylated Human 2B4, Fc,Avitag with a linear range of 0,01-0,156 μg/ml (QC tested)
  • Gene ID:
    Q9BZW8-2
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    NAIL|CD244|SLAMF4|h2B4|2B4|NKR2B4
  • Protein/peptide name:
    2B4
  • Purity:
    >95%
  • Molecular weight:
    50,7 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item