Human recombinant IL-12B (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Human IL-12B, Fc Tag (Cat. No. NK2-H5258) at 5 μg/ml (100 μl/well) can bind Biotinylated Human IL-12 R beta 1, Fc,Avitag with a linear range of 5 to 40 ng/ml (QC tested).
- Gene ID:P29460
- Protein synonyms:NKSF|CLMF|CLMF2|NKSF2|p40|IL12B
- Protein/peptide name:IL-12B
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:61,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together