Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant Her2 (from HEK293 cells)
Human recombinant Her2 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant Her2 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Mouse IgG2a Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Trastuzumab at 0,2 μg/ml (100 μl/well) can bind Human Her2, Mouse IgG2a Fc Tag with a linear range of 0,3 to 10 ng/ml (QC tested).
  • Gene ID:
    P04626-1
  • Protein synonyms:
    NEU|MLN19|TKR1|ERBB2|CD340|HER2|HER-2/neu|NGL
  • Protein/peptide name:
    Her2
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    96,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together