Human recombinant SLC1A5 (from E.coli)
Human SLC1A5 Protein Tag, TrxA Tag.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:TrxA Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Gene ID:Q15758-1
- Protein synonyms:RDRC|RD114|ASCT2M7VS1|AAAT|RDR|R16|RDRCFLJ31068|ATB(0)|ASCT2|M7V1ATBO|M7V1
- Protein/peptide name:SLC1A5
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:22,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Frequently Bought Together