Order Entry
Netherlands
ContactUsLinkComponent
Rabbit recombinant CTLA-4 (from HEK293)
Rabbit recombinant CTLA-4 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Rabbit recombinant CTLA-4 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rabbit
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised human B7-1, mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBB71-H52A4) at 5 μg/ml (100 μl/well)can bind Rabbit CTLA-4, Fc Tag (Cat. No. ACRBCT4-R5250) with a linear range of 0,1 - 0,8 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised human B7-2, His Tag (Cat. No. ACRBCD6-H5223) at 5 μg/ml (100 μl/well)can bind rabbit CTLA-4, Fc Tag (Cat. No. ACRBCT4-R5250) with a linear range of 5 - 78 ng/ml (Routinely tested).
  • Gene ID:
    P42072
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CTLA4|CD152
  • Protein/peptide name:
    CTLA-4
  • Purity:
    >95%
  • Molecular weight:
    40,3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together