Mouse recombinant OX40 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised mouse OX40 ligand, His Tag (Cat. No. ACRBTN4-M5241) at 2 μg/ml (100 μl/well) can bind biotinylated mouse OX40, His, Avitag (Cat. No. ACRBOX0-M82E5) with a linear range of 0,6 -10 ng/ml (QC tested).
- Gene ID:P47741-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TXGP1L|ACT35|OX40|TNFRSF4|OX40L receptor|CD134
- Protein/peptide name:OX40
- Purity:>90%
- Molecular weight:25,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together