Order Entry
Netherlands
ContactUsLinkComponent
Mouse recombinant glypican 3 (from HEK293)
Mouse recombinant glypican 3 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse recombinant glypican 3 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised human FGF basic, tag free (Cat. No. ACRBBFF-H4117) at 0,5 μg/ml (100 μl/well) can bind mouse glypican 3, His tag (Cat. No. ACRBGP3-M52H3) with a linear range of 1 - 16 ng/ml (QC tested).
  • Gene ID:
    Q8CFZ4-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    GPC3|SGBS|SGB|Glypican-3|SDYS |MXR7|SGBS1|OCI5|GTR2-2|DGSX
  • Protein/peptide name:
    Glypican 3
  • Purity:
    >95%
  • Molecular weight:
    38,0 kDa and 24,4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together