Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant CD28H (from HEK293)
Human recombinant CD28H (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant CD28H (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human B7-H7, His Tag (Cat. No. B77-H52H5) at 5 μg/ml (100 μl/well) can bind Human CD28H, Fc Tag (Cat. No. CDH-H5251) with a linear range of 1 - 20 ng/ml (QC tested).
  • Gene ID:
    Q96BF3-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    TMIGD2|IGPR1
  • Protein/peptide name:
    CD28H
  • Purity:
    >95%
  • Molecular weight:
    40,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together