Human recombinant CD27 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human CD27 Ligand, Fc Tag (Cat. No. TN7-H526x) at 1 μg/ml (100 μl/well) can bind Human CD27, Mouse IgG2a Fc Tag (Cat. No. CD7-H5257) with a linear range of 0,005 - 0,078 μg/ml (QC tested). Flow Cytometry assay shows that Human CD27, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-H5257) can bind to Raji cells Surface Human CD70. The concentration of CD27 used is 10 μg/ml (Routinely tested).
- Gene ID:P26842-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Tp55|S152|T14|TNFRSF7|CD27
- Protein/peptide name:CD27
- Purity:>95%
- Molecular weight:46,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Specifications
Frequently Bought Together