Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant CD96 (from HEK293)
Human recombinant CD96 (from HEK293)
Catalog # ACRBTAE-H52H0-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human recombinant CD96 (from HEK293)
Catalog # ACRBTAE-H52H0-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Seq:
    02
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Gene ID:
    P40200-2
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    TACTILE
  • Protein/peptide name:
    CD96
  • Purity:
    >90%
  • Molecular weight:
    55,4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

About this item