Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant neuropilin-1 (from HEK293)
Human recombinant neuropilin-1 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant neuropilin-1 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation,; Measured by its binding ability in a functional ELISA, Immobilised ActiveMax® Human VEGF165, Tag Free (HPLC-verified) (Cat, No, VE5-H4210) at 2 μg/ml (100 μl/well) can bind Biotinylated Human Neuropilin-1, His,Avitag (Cat, No, NR1-H82E3) with a linear range of 0,5-16 ng/mL (QC tested).
  • Gene ID:
    O14786-1 (V179A)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    NRP1|Neuropilin-1|VEGF165R|NRP|CD304
  • Protein/peptide name:
    Neuropilin-1
  • Purity:
    >95%
  • Molecular weight:
    73,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together