2-(6-Ethoxy-2-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₅H₂₂BFO₃
MDL Number:MFCD32707324
:280.143 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together