
2,4-Dibromo-5-(trifluoromethyl)-1H-imidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:81654-02-6
:
Formula:C₄HBr₂F₃N₂
MDL Number:MFCD08458161
:293.867 g/mol
Melting Pt:162…164 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together