

Methyl 3-methoxy-2,3,3-trifluoropropanoate 93%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:758-69-0
:
Formula:C₅H₇F₃O₃
MDL Number:MFCD07784247
:172.102 g/mol
Boiling Pt:90…92 °C (50 mm Hg)
Density:1,301 g/cm³ (25 °C)
Storage Temperature:Kamertemperatuur
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together