
(R)-6-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:731859-02-2
:
Formula:C₉H₁₀FN·ClH
MDL Number:MFCD16295032
:187.642 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together