
1-(2-(Trifluoromethyl)pyridin-4-yl)ethanone 97%
Catalog # APOSPC902889-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:189.135 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1060810-86-7
Formula:C₈H₆F₃NO
MDL Number:MFCD11847720