Order Entry
Netherlands
ContactUsLinkComponent
2-Ethoxy-1-fluoro-4-nitrobenzene 97%
2-Ethoxy-1-fluoro-4-nitrobenzene 97%
Catalog # APOSPC902093-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Ethoxy-1-fluoro-4-nitrobenzene 97%
Catalog # APOSPC902093-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :185.152 g/mol
Storage Temperature:Ambient
CAS Number:1093656-34-8
Formula:C₈H₈FNO₃
MDL Number:MFCD20133797