Ethyl 4-({[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}carbonyl)-5-methylisoxazole-3-carboxylate 97%
APOSPC8477-100MG
:
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
:
:377.703 g/mol
:94…96 °C
:Ambient
:338397-35-6
:C₁₄H₁₁ClF₃N₃O₄
:MFCD00664738