Diethyl 3-{[3-(trifluoromethyl)phenyl]carbamoyl}-1,2-cyclopropanedicarboxylate 97%
Catalog # APOSPC8454-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:373.324 g/mol
Melting Pt:115…117 °C
Storage Temperature:Ambient
CAS Number:318237-96-6
Formula:C₁₇H₁₈F₃NO₅
MDL Number:MFCD01315628