
Ethyl 2-chloro-4-fluorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:167758-87-4
:
Formula:C₉H₈ClFO₂
MDL Number:MFCD06204388
:202.61 g/mol
Boiling Pt:239…241 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together