Order Entry
Netherlands
ContactUsLinkComponent
1-[2,6-Bis(trifluoromethyl)quinol-4-yl]piperazine 97%
Irritant
1-[2,6-Bis(trifluoromethyl)quinol-4-yl]piperazine 97%
Catalog # APOSPC7615-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-[2,6-Bis(trifluoromethyl)quinol-4-yl]piperazine 97%
Catalog # APOSPC7615-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :349.274 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:401567-74-6
Formula:C₁₅H₁₃F₆N₃
MDL Number:MFCD03094433