Order Entry
Netherlands
ContactUsLinkComponent
5-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulphonyl]thiophene-2-sulphonyl chloride
5-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulphonyl]thiophene-2-sulphonyl chloride
Catalog # APOSPC6954-100MG
Supplier:  Apollo Scientific
CAS Number:  
5-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulphonyl]thiophene-2-sulphonyl chloride
Catalog # APOSPC6954-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :426.239 g/mol
Melting Pt:126…127 °C
Storage Temperature:Ambient
CAS Number:175203-00-6
Formula:C₁₀H₄Cl₂F₃NO₄S₃
MDL Number:MFCD00068006