
1H,1H-Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecan-1-ol 97%
Catalog # APOSPC6603-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:648.113 g/mol
Boiling Pt:70 °C (1 mm Hg)
Density:1.7595 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:14620-81-6
Formula:C₁₂H₃F₂₃O₄
MDL Number:MFCD00069091