
5-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]thiophene-2-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175202-29-6
:
Formula:C₁₀H₇F₃N₂O₂S
MDL Number:MFCD00085143
:276.235 g/mol
Melting Pt:213…218 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together