Lithium perfluoro(2,6-dimethyl-5-oxaheptane 2-oxide)
Catalog # APOSPC6468-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:457.998 g/mol
Boiling Pt:155…157 °C (0.3 mm Hg)
Flash Pt:>110 °C
Storage Temperature:Ambient
CAS Number:78740-39-3
Formula:C₈F₁₇O₂·Li
MDL Number:MFCD10567150