
5-[5-(Trifluoromethyl)pyridin-2-yl]-1H-tetrazole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175334-70-0
:
Formula:C₇H₄F₃N₅
MDL Number:MFCD00205782
:215.135 g/mol
Melting Pt:218…222 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together