[5-[(4-Chlorophenyl)sulphonyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:318469-34-0
:
Formula:C₁₂H₁₀ClF₃N₂O₃S
MDL Number:MFCD00172660
:354.733 g/mol
Melting Pt:103…105 °C
Storage Temperature:Ambient
Frequently Bought Together