5-Amino-2-(methylsulphonyl)benzotrifluoride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:252561-92-5
:
Formula:C₈H₈F₃NO₂S
MDL Number:MFCD12807191
:239.215 g/mol
Melting Pt:141…141 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together