
4-(Trifluoromethyl)benzothiophene-2-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:908566-20-1
:
Formula:C₁₀H₅F₃OS
MDL Number:MFCD27932089
:230.206 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together