
4-(Trifluoromethyl)benzothiophene-2-carbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2115333-67-8
:
Formula:C₁₀H₄F₃NS
MDL Number:MFCD31567871
:227.206 g/mol
Melting Pt:85…87 °C
:GHS07
Storage Temperature:Kamertemperatuur
Frequently Bought Together