
4-Bromo-2,6-difluorobenzenesulphonamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:263349-74-2
:
Formula:C₆H₄BrF₂NO₂S
MDL Number:MFCD06409103
:272.067 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together