

3-(Trifluoromethyl)phenyl chloroformate 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:95668-29-4
:
Formula:C₈H₄ClF₃O₂
MDL Number:MFCD02093719
:224.564 g/mol
UN:3277
ADR:6.1+8,II
:GHS05
:GHS06
Storage Temperature:Refrigerator
Frequently Bought Together