1-(4-Bromophenyl)-2,2,2-trifluoroethanol ≥96%
Catalog # APOSPC540041-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Verpakking:Bottle
- VPE:250 mg
- Seq:2590
Specifications
About this item
:
:255.032 g/mol
Storage Temperature:Koelkast
CAS Number:76911-73-4
Formula:C₈H₆BrF₃O
MDL Number:MFCD12786801