Order Entry
Netherlands
ContactUsLinkComponent
(S)-1-(5-Fluoropyrimidin-2-yl)ethanamine hydrochloride 98%
(S)-1-(5-Fluoropyrimidin-2-yl)ethanamine hydrochloride 98%
Catalog # APOSPC540007-250MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-1-(5-Fluoropyrimidin-2-yl)ethanamine hydrochloride 98%
Catalog # APOSPC540007-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :177.607 g/mol
Storage Temperature:Ambient
CAS Number:935667-21-3
Formula:C₆H₈FN₃·ClH
MDL Number:MFCD18207119