Order Entry
Netherlands
ContactUsLinkComponent
3-Chloro-1-hydroxy-5-(trifluoromethyl)pyridin-2(1H)-one
Irritant
3-Chloro-1-hydroxy-5-(trifluoromethyl)pyridin-2(1H)-one
Catalog # APOSPC53581-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Chloro-1-hydroxy-5-(trifluoromethyl)pyridin-2(1H)-one
Catalog # APOSPC53581-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :213.542 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1823183-80-7
Formula:C₆H₃ClF₃NO₂
MDL Number:MFCD29042535