
Methyl 2,4-Difluoro-5-(trifluoromethyl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1823296-98-5
:
Formula:C₉H₅F₅O₂
MDL Number:MFCD22393602
:240.127 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together