
Methyl 4-(pentafluoroethyl)-3-(trifluoromethyl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1980063-30-6
:
Formula:C₁₁H₆F₈O₂
MDL Number:MFCD30062865
:322.151 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together