Order Entry
Netherlands
ContactUsLinkComponent
2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene 95%
2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene 95%
Catalog # APOSPC520590-10G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-1-(difluoromethoxy)-4-(trifluoromethyl)benzene 95%
Catalog # APOSPC520590-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :291.013 g/mol
Storage Temperature:Ambient
CAS Number:954236-03-4
Formula:C₈H₄BrF₅O
MDL Number:MFCD11036074