
2-Bromo-6-methyl-4-nitrophenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1403666-47-6
:
Formula:C₈H₅BrF₃NO₅S
MDL Number:MFCD27975431
:364.093 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together