1-[6-(Trifluoromethyl)-2-pyridyl]cyclopropanecarbonitrile
Catalog # APOSPC520379-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.171 g/mol
Storage Temperature:Ambient
CAS Number:1346544-42-0
Formula:C₁₀H₇F₃N₂
MDL Number:MFCD23134904